missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-FBXL3 (Ser320) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567503
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats and is localized in the nucleus.
Specifications
| Phospho-FBXL3 (Ser320) | |
| Polyclonal | |
| Unconjugated | |
| FBXL3 | |
| Afh; AU041772; AW212966; FBK; FBL3; Fbl3a; F-box and leucine rich repeat protein 3; F-box and leucine-rich repeat protein 3; F-box and leucine-rich repeat protein 3A; F-box protein Fbl3a; F-box/LRR-repeat protein 3; F-box/LRR-repeat protein 3A; FBXL3; Fbxl3a; Ovtm; Play68; Protein after-hours; protein overtime | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26224, 306129, 50789 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C4V4, Q9UKT7 | |
| FBXL3 | |
| Synthesized peptide sequence derived from Human FBXL3 around phosphorylation site of serine 320 (S-V-S(p)-K-D). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction