missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-FANCG (Ser383) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538154
This item is not returnable.
View return policy
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group G.
Specifications
| Phospho-FANCG (Ser383) | |
| Polyclonal | |
| Unconjugated | |
| FANCG | |
| Fanconi anemia group G protein, Protein FACG, DNA repair protein XRCC9, FANCG, XRCC9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2189 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15287 | |
| FANCG | |
| A synthetic phosphopeptide derived from human FANCG around the phosphorylation site of Ser383 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction