missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Ezrin (Thr567) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15544701
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15544701 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544701 Supplier Invitrogen™ Supplier No. PA537763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is MDA cells.

The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Ezrin (Thr567)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ezr
Gene Accession No. P15311, P26040, P31977
Gene Alias AW146364; CVIL; CVL; Cytovillin; cytovillin 2; DKFZp762H157; epididymis secretory protein Li 105; ezr; ezr.L; ezr.S; EZRI; Ezrin; ezrin L homeolog; ezrin L homeolog; ezrin S homeolog; FLJ26216; HEL-S-105; MGC1584; p81; R75297; VIL2; villin 2; villin 2 (ezrin); villin-2; XELAEV_18027043mg
Gene Symbols Ezr
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of threonine 567 (Y-K-T(p)-L-R) derived from Human Ezrin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22350, 54319, 7430
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.