missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Ezrin/Radixin/Moesin (Thr558) Polyclonal Antibody, Invitrogen™

Product Code. 15595591
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15595591 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15595591 Supplier Invitrogen Supplier No. PA538679

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-Ezrin/Radixin/Moesin (Thr558) Polyclonal antibody specifically detects Phospho-Ezrin/Radixin/Moesin (Thr558) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Ezrin/Radixin/Moesin (Thr558)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MSN
Gene Accession No. O35763, P15311, P26038, P26040, P26041, P26043, P31977, P35241
Gene Alias AA516625; AW146364; C78546; CVIL; CVL; cytovillin; cytovillin 2; DFNB24; epididymis luminal protein 70; epididymis secretory protein Li 105; ESP10; EZR; Ezrin; HEL70; HEL-S-105; membrane-organizing extension spike protein; Moesin; MSN; p81; R75297; radixin; radixin isoform; RDX; VIL2; villin 2; villin 2 (ezrin); villin-2
Gene Symbols Ezr, MSN, RDX
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human Moesin/Ezrin/Radixin around the phosphorylation site of Thr558 (Y-K-TP-L-R)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17698, 19684, 22350, 315655, 4478, 54319, 5962, 7430, 81521
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.