missing translation for 'onlineSavingsMsg'
Learn More

Phospho-ETK (Tyr40) Polyclonal Antibody, Invitrogen™

Product Code. 15505291
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15505291 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15505291 Supplier Invitrogen Supplier No. PA538363

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ETK, also known as BMX, is a member of the Bruton′s tyrosine kinase (Btk) family. It is expressed in a variety of hematopoietic, epithelial and endothelial cells. Etk, like other Btk family members, contains a pleckstrin homology (PH) domain and Src homology SH3 and SH2 domains. It participates in multiple signal transduction pathways. Phosphorylation of Tyr566 by Src kinase is required for activation of Etk in vivo. In endothelial and epithelial cells, Etk is regulated by FAK through phosphorylation at Tyr40.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ETK (Tyr40)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BMX
Gene Accession No. P51813, P97504
Gene Alias Cytoplasmic tyrosine-protein kinase BMX, Bone marrow tyrosine kinase gene in chromosome X protein, Epithelial and endothelial tyrosine kinase, NTK38, BMX, ETK
Gene Symbols BMX
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr40 (L-S-YP-Y-E)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12169, 660
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.