Learn More
Invitrogen™ Phospho-ERK1/2 (Thr202, Tyr204) Monoclonal Antibody (MILAN8R), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£107.00 - £276.00
Specifications
| Antigen | Phospho-ERK1/2 (Thr202, Tyr204) |
|---|---|
| Clone | MILAN8R |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15548236
|
Invitrogen™
17-9109-41 |
25 Tests |
£107.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15370408
![]() |
Invitrogen™
17-9109-42 |
100 Tests |
£276.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This MILAN8R monoclonal antibody recognizes human and mouse extracellular signal-regulated kinases 1 and 2 (also known as ERK1/2, p44/p42, or MAPK3/1) when phosphorylated on T202/Y204. ERK1/2 belong to a family of conserved serine/threonine protein kinases known as mitogen-activated protein kinases (MAPKs) that are involved in many cellular programs such as proliferation, differentiation, motility, and survival. ERK1/2 signaling is activated in response to numerous extracellular stimuli including mitogens, growth factors, and cytokines. The primary activators of ERK1/2 are MEK1 and MEK2 which act by phosphorylating the activation loop residues T202/Y204 and T185/Y187 in ERK1 and ERK2, respectively. Several downstream targets of ERK1/2 have been identified, including p90RSK and the transcription factor Elk-1. ERK1/2 are negatively regulated by MAPK phosphatases, known as DUSPs or MKPs, as well as by chemical inhibitors of MEK including U0126 and PD98059. Disruption of the ERK pathway is common in many types of cancer. Specificity of this MILAN8R clone was determined by ELISA, flow cytometry, and western blotting. Applications Reported: This MILAN8R antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MILAN8R antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells.
Phospho-ERK1/2 (Thr202, Tyr204) Monoclonal antibody specifically detects Phospho-ERK1/2 (Thr202, Tyr204) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| Phospho-ERK1/2 (Thr202, Tyr204) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupII; HS44KDAP; HUMKER1A; I79_009500; I79_018350; insulin-stimulated MAP2 kinase; l(2)41Ac; l(2R)EMS45-39; M phase MAP kinase; MAP kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MapK; MAP-k; MAPK 1; MAPK 2; MAPK 3; MAPK1; mapk1.S; mapk1a; mapk1-a; mapk1-b; mapk2; mapk3; MAP-kinase; MBP kinase; Microtubule-associated protein 2 kinase; mitogen activated protein kinase; mitogen activated protein kinase 1; mitogen activated protein kinase 3; mitogen-activated 3; mitogen-activated protein kinase; mitogen-activated protein kinase 1; mitogen-activated protein kinase 1 S homeolog; mitogen-activated protein kinase 1a; mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; Mitogen-activated protein kinase ERK-A; MNK1; mpk1; Mtap2k; Myelin basic protein kinase; Myelin xP42 protein kinase; p38; p40; p41; p41mapk; p42; p42 MAP Kinase; P42MAPK; p42-MAPK; p44; p44 MAP kinase; p44erk1; p44-ERK1; p44mapk; p44-MAPK; pERK; p-ERK; phospho-ERK; pMapK; pp42/MAP kinase; Prkm1; PRKM2; Prkm3; protein kinase; Protein rolled; protein tyrosine kinase ERK2; RE08694p; RL; rl/ERK; rl/MAPK; rll; rl-PA; rl-PC; rl-PD; rl-PE; rl-PG; rl-PH; rl-PI; roll; rolled; sem; sevenmaker; SR2-1; Su(Raf)2B; wu:fi06b09; XELAEV_18010534mg; xp42; zERK1; zERK2 | |
| MAPK1, MAPK3 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| MILAN8R | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human, Mouse | |
| P27361, P28482, P63085, Q63844 | |
| 26413, 26417, 5594, 5595 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.