missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ERK1/ERK2 (Phospho) [pT202/pY204]/[pT185/pY187] Multispecies InstantOne™ ELISA Kit

InstantOne ELISA

Brand:  Invitrogen™ 85-86012-253

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15550767

  • £11490.00 / Pack of 25
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Alert:

Please contact Technical Service for a lot specific COA


Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Capture Antibody Reagent
phospho-ERK 1/2 (Thr202/Tyr204, Thr185/Tyr187) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate seal

Description

Description

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 in cell lysates. Detection of rat ERK 1/2 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Specifications

Specifications

Hamster, Human, Mouse
25 x 384 Tests
p44/p42, MAP Kinase 1/2
ELISA
2°C to 8°C
ERK 1, ERK 2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only