missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EpoR (Tyr454, Tyr456) Recombinant Superclonal™ Antibody (3HCLC)
Phospho-EpoR (Tyr454, Tyr456) Recombinant Polyclonal Antibody (3HCLC)
Brand: Invitrogen 710484
This item is not returnable.
View return policy
Description
Phospho-EpoR (Tyr454, Tyr456) Recombinant Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-EpoR (Tyr454, Tyr456) | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P19235 | |
| EPOR | |
| Phosphorylation of the human EPO Receptor at the Tyr454 and Tyr456. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 3HCLC | |
| Unconjugated | |
| EPOR | |
| EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction