missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EpoR (Tyr368) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538483
This item is not returnable.
View return policy
Description
Receptor for erythropoietin. Mediates erythropoietin- induced erythroblast proliferation and differentiation. Upon EPO stimulation, EPOR dimerizes triggering the JAK2/STAT5 signaling cascade. In some cell types, can also activate STAT1 and STAT3. May also activate the LYN tyrosine kinase.
Specifications
| Phospho-EpoR (Tyr368) | |
| Polyclonal | |
| Unconjugated | |
| EPOR | |
| EPO R, EPO-R, epor, pEpoR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13857, 2057, 24336 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19235, P14753, Q07303 | |
| EPOR | |
| A synthetic phosphopeptide derived from human Epo-R around the phosphorylation site of Tyr368 (D-T-YP-L-V) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction