missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Ephrin-B (Mouse/Rat Y343) Antibody, R&D Systems™

Product Code. 18048131
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18048131 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18048131 Supplier R&D Systems Supplier No. PPS049
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide columns.

Ephrin-B Polyclonal specifically detects Ephrin-B in Human, Mouse, Rat, Chicken, Xenopus samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin-B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias craniofrontonasal syndrome (craniofrontonasal dysplasia), EFL3, EFL-3, ELK ligand, ELK-L, EPH-related receptor tyrosine kinase ligand 2, EphrinB, ephrin-B1, EPLG2CFNS, LERK-2, LERK2CFND, ligand of eph-related kinase 2, MGC8782
Gene Symbols EFNB1
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phospho-Y331 of chicken Ephrin-B
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1947
Test Specificity This antibody is specific for the ∼45 kDa Ephrin-B phosphorylated at Y343 in Western blots of rat testis.
Target Species Human, Mouse, Rat, Chicken, Xenopus species
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.