missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EphA8 (Tyr839) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105876
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of EPHA8 only when phosphorylated at Tyr839.
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
Specifications
| Phospho-EphA8 (Tyr839) | |
| Polyclonal | |
| Unconjugated | |
| Epha8 | |
| AW047546; Eek; EK3; epa8; EPH- and ELK-related kinase; EPH- and ELK-related tyrosine kinase; Eph receptor A8; Epha8; EPH-like kinase 3; Ephrin type-A receptor 8; Ephrin type-A receptor 8 precursor (Tyrosine-protein kinase receptor EEK) (EPH-and ELK-related kinase); Hek3; hydroxyaryl-protein kinase; KIAA1459; mKIAA1459; protein-tyrosine kinase; tyrosine-protein kinase receptor EEK; tyrosylprotein kinase | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 13842, 2046, 60589 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09127, P29321, P29322 | |
| Epha8 | |
| A synthesized peptide derived from human EPHA8(Accession P29322), corresponding to amino acid residues around phosphorylated Tyr839. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction