missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Epha3 (Tyr779) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597390
This item is not returnable.
View return policy
Description
Immunogen sequence: AAYTT; Positive Samples: 293T, Hela; Cellular Location: Cell membrane, Single-pass type I membrane protein.
EPHA3 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA3 encodes a protein that binds ephrin-A ligands and receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.
Specifications
| Phospho-Epha3 (Tyr779) | |
| Polyclonal | |
| Unconjugated | |
| EPHA3 | |
| AW492086; Cek4; EK4; End3; EPH receptor A3; EPHA3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; Hek; Hek4; Human embryo kinase; human embryo kinase 1; I79_002835; Mek4; rEK4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase TYRO4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2042 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P29320 | |
| EPHA3 | |
| A synthetic phosphorylated peptide around Y779 of human EphA3 (NP_0052242). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction