missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EPB41 (Tyr660) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540232
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal dominant, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4. 1. 'Rh-unlinked' forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.
Specifications
| Phospho-EPB41 (Tyr660) | |
| Polyclonal | |
| Unconjugated | |
| EPB41 | |
| 4.1R; AI415518; Band 4.1; D4Ertd442e; E41P; EL1; elliptocytosis 1, RH-linked; Elp1; Elp-1; EPB4.1; EPB41; erythrocyte membrane protein band 4.1; erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked); erythrocyte protein band 4.1; erythrocyte skeletal protein; erythrocyte surface protein band 4.1; HE; Kiaa4056; mKIAA4056; P4.1; protein 4.1; Protein 4.1R; RGD1564762 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2035, 269587 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P11171, P48193 | |
| EPB41 | |
| Synthesized peptide derived from human 4.1R around the phosphorylation site of Y660. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction