missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41 Polyclonal Antibody

Phospho-EPB41 (Tyr418, Tyr660) Polyclonal Antibody
Brand: Invitrogen PA5102613
This item is not returnable.
View return policy
Description
Phospho-EPB41 (Tyr418, Tyr660) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| EPB41 | |
| Polyclonal | |
| Unconjugated | |
| EPB41 | |
| 4.1R; AI415518; Band 4.1; D4Ertd442e; E41P; EL1; elliptocytosis 1, RH-linked; Elp1; Elp-1; EPB4.1; EPB41; erythrocyte membrane protein band 4.1; erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked); erythrocyte protein band 4.1; erythrocyte skeletal protein; erythrocyte surface protein band 4.1; HE; Kiaa4056; mKIAA4056; P4.1; protein 4.1; Protein 4.1R; RGD1564762 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2035, 269587, 313052 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11171, P48193 | |
| EPB41 | |
| A synthesized peptide derived from human EPB41(Accession P11171), corresponding to amino acid residues H681-L731. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction