missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-eNOS (Ser1177) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517917
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains.
Specifications
| Phospho-eNOS (Ser1177) | |
| Polyclonal | |
| Unconjugated | |
| NOS3 | |
| 2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 287024, 397557, 4846 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 79 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P29473, P29474, Q28969 | |
| NOS3 | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer1177 of human eNOS. | |
| 100 μL | |
| Primary | |
| Bovine, Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction