missing translation for 'onlineSavingsMsg'
Learn More

Phospho(enol)pyruvic Acid Monopotassium Salt, TRC

Product Code. 30426366
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30426366 100 mg
30421016 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426366 Supplier Toronto Research Chemicals Supplier No. TRCP360870100MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-07-0
Molecular Formula C3 H4 O6 P . K
Molecular Weight (g/mol) 206.132
Synonym 2-Propenoic acid, 2-(phosphonooxy)-, potassium salt (1:1), 2-Propenoic acid, 2-(phosphonooxy)-, monopotassium salt (9CI), Acrylic acid, 2-hydroxy-, dihydrogen phosphate, monopotassium salt (8CI), Monopotassium phosphoenolpyruvate, Phosphoenolpyruvic acid potassium salt, 2-(Phosphonooxy)-2-propenoic acid monopotassium salt, Phosphoenolpyruvic acid monopotassium salt, Phospho(enol)pyruvic acid monopotassium salt
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
SMILES [K+].OC(=O)C(=C)OP(=O)(O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H5O6P.K/c1-2(3(4)5)9-10(6,7)8;/h1H2,(H,4,5)(H2,6,7,8);/q;+1/p-1
Formula Weight 205.938
Chemical Name or Material Phospho(enol)pyruvic Acid Monopotassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.