missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-eIF4E (Ser209) Recombinant Rabbit Monoclonal Antibody (0I0D10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555315
This item is not returnable.
View return policy
Description
Phospho-eIF4E (Ser209) Recombinant Monoclonal Antibody for Western Blot, ELISA
eIF4E is involved in several cellular processes including enhanced translational efficiency, splicing, mRNA stability, and RNA nuclear export. All eukaryotic cellular mRNAs are blocked at their 5-prime ends with the 7-methylguanosine cap structure, m7GpppX (where X is any nucleotide). EIF4E is a eukaryotic translation initiation factor involved in directing ribosomes to the cap structure of mRNAs. It is a 24-kD polypeptide that exists as both a free form and as part of a multiprotein complex termed EIF4F. The EIF4E polypeptide is the rate-limiting component of the eukaryotic translation apparatus and is involved in the mRNA-ribosome binding step of eukaryotic protein synthesis. The other subunits of EIF4F are a 50-kD polypeptide, termed EIF4A, that possesses ATPase and RNA helicase activities, and a 220-kD polypeptide, EIF4G.
Specifications
| Phospho-eIF4E (Ser209) | |
| Recombinant Monoclonal | |
| 0.75 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06730, P63074 | |
| Eif4e | |
| A phospho specific peptide corresponding to residues surrounding S209 of human Phospho-eIF4E-S209. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0I0D10 | |
| Unconjugated | |
| Eif4e | |
| AUTS19; CBP; EG668879; EIF4E; EIF-4E; EIF4E1; EIF4EL1; Eif4e-ps; EIF4F; eIF-4F 25 kDa subunit; eukaryotic translation initiation factor 4E; eukaryotic translation initiation factor 4E-like 1; eukaryotic translation initiation factor 4E-like protein; eukaryotic translation initiation factor 4-like protein; If4e; MGC111573; mRNA cap-binding protein; OTTHUMP00000219703; OTTHUMP00000219704; OTTHUMP00000219705; translation initiation factor eIF-4E | |
| Rabbit | |
| Protein A | |
| RUO | |
| 117045, 1977 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction