missing translation for 'onlineSavingsMsg'
Learn More

Phospho-EIF2S1 (Ser51) Mouse Monoclonal Antibody, Proteintech

Product Code. 30236497 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30236497 100 μL 100µL
30233338 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30236497 Supplier Proteintech Supplier No. 680231IG100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Phospho-EIF2S1 (Ser51) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-EIF2S1 (Ser51). This antibody recognizes human, mouse, rat antigen. The Phospho-EIF2S1 (Ser51) antibody has been validated for the following applications: WB, IF/ICC, FC (Intra), ELISA analysis.

Specifications

Antigen EIF2S1 (Ser51)
Applications Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
Classification Monoclonal
Clone 1A4A11
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A
Host Species Mouse
Immunogen Peptide
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1965
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 36 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.