missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EIF2S1 (Ser51) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68023-1-IG-20UL
This item is not returnable.
View return policy
Description
The Phospho-EIF2S1 (Ser51) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-EIF2S1 (Ser51). This antibody recognizes human, mouse, rat antigen. The Phospho-EIF2S1 (Ser51) antibody has been validated for the following applications: WB, IF/ICC, FC (Intra), ELISA analysis.Specifications
| EIF2S1 (Ser51) | |
| Monoclonal | |
| 1000 μg/mL | |
| EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 36 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 1A4A11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 1965 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction