missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-eIF2 beta (Ser67) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118774
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of EIF2S2 only when phosphorylated at Ser67.
Eukaryotic translation initiation factor 2 (EIF-2) functions in the early steps of protein synthesis by forming a ternary complex with GTP and initiator tRNA and binding to a 40S ribosomal subunit. EIF-2 is composed of three subunits, alpha, beta, and gamma, with the protein encoded by this gene representing the beta subunit. The beta subunit catalyzes the exchange of GDP for GTP, which recycles the EIF-2 complex for another round of initiation. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| Phospho-eIF2 beta (Ser67) | |
| Polyclonal | |
| Unconjugated | |
| EIF2S2 | |
| 2810026E11Rik; 38kDa; AA408636; AA571381; AA986487; AW822225; D2Ertd303e; EIF2; EIF2B; EIF2BA; EIF2beta; eIF-2-beta; EIF2S2; Eukaryotic translation initiation factor 2 subunit 2; eukaryotic translation initiation factor 2 subunit beta; eukaryotic translation initiation factor 2, subunit 2 (beta); eukaryotic translation initiation factor 2, subunit 2 (beta, 38kDa); eukaryotic translation initiation factor 2, subunit 2 beta; eukaryotic translation initiation factor 2, subunit 2 beta, 38kDa; PPP1R67; protein phosphatase 1, regulatory subunit 67 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 67204, 8894 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20042, Q99L45 | |
| EIF2S2 | |
| A synthesized peptide derived from human EIF2S2(Accession P20042), corresponding to amino acid residues around phosphorylated Ser67. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction