missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EEF2 (Thr56) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517839
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
Specifications
| Phospho-EEF2 (Thr56) | |
| Polyclonal | |
| Unconjugated | |
| EEF2 | |
| EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100689051, 13629, 1938, 29565, 396325 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 84 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P05197, P09445, P13639, P58252, Q90705 | |
| EEF2 | |
| Synthetic phosphopeptide corresponding to residues surrounding pThr56 of human eEF2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Hamster, Chicken | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction