Learn More
Invitrogen™ Phospho-EEF1A2 (Ser358) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA535404
Product Code. 15318174
  • £439.00 / 100µL
Description
This antibody is predicted to react with chicken, non-human primate, rat, sheep and Xenopus based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.
This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. [provided by RefSeq, Mar 2014].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-EEF1A2 (Ser358)
Polyclonal
Unconjugated
EEF1A2
Eef1a; EEF1A2; eEF1A-2; EEF1AL; EF1A; EF-1-alpha-2; EIEE33; elongation factor 1 A-2; elongation factor 1-alpha 2; Eukaryotic elongation factor 1 A-2; eukaryotic translation elongation factor 1 alpha 2; HS1; MRD38; Ps10; RATPS10; S1; statin S1; Statin-like protein; statin-S1; STN; Stnl; wasted; wst
Rabbit
Affinity chromatography
RUO
13628, 1917
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.1 mg/mL
0.01M HEPES with 0.1mg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
P62631, Q05639
EEF1A2
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser358 of human eEF1A2 conjugated to KLH.
100 μL
Primary
Human, Mouse