missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DRP1 (Ser616) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564821
This item is not returnable.
View return policy
Description
Phospho-DRP1 (Ser616) Polyclonal Antibody detects endogenous levels of DRP1 only when phosphorylated at Ser616.
A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
Specifications
| Phospho-DRP1 (Ser616) | |
| Polyclonal | |
| Unconjugated | |
| DNM1L | |
| 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10059, 114114, 74006 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| O00429, O35303, Q8K1M6 | |
| DNM1L | |
| A synthesized peptide derived from human DRP1 (Phospho-Ser616). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction