missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DOK2 (Tyr345) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118727
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of DOK2 only when phosphorylated at Tyr345.
The protein encoded by this gene is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. This encoded protein binds p120 from CML cells.
Specifications
| Phospho-DOK2 (Tyr345) | |
| Polyclonal | |
| Unconjugated | |
| DOK2 | |
| docking protein 2; docking protein 2, 56kD; docking protein 2, 56kDa; Dok2; Dok-2; DokR; Dok-R; dok-related protein; Downstream of tyrosine kinase 2; Frip; IL-Four Receptor Interacting Protein; IL-four receptor-interacting protein; LOW QUALITY PROTEIN: docking protein 2; docking protein 2; p56 dok-2; p56(dok-2); p56DOK; p56dok-2 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 13449, 9046 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60496, O70469 | |
| DOK2 | |
| A synthesized peptide derived from human DOK2(Accession O60496), corresponding to amino acid residues around phosphorylated Tyr345. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction