missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-DOK1 (Ser450) Polyclonal Antibody

Phospho-DOK1 (Ser450) Polyclonal Antibody
Brand: Invitrogen PA5143682
This item is not returnable.
View return policy
Description
Phospho-DOK1 (Ser450) Polyclonal Antibody for Western Blot
Specifications
| Phospho-DOK1 (Ser450) | |
| Polyclonal | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P97465, Q4QQV2, Q99704 | |
| DOK1 | |
| Phospho-Dok1 (Ser-450) synthetic peptide (coupled to KLH) corresponds to amino acids surrounding serine 450 in human Dok1. This sequence is conserved in Dok1 from rat (Ser-449) and mouse (Ser-451). The site is not conserved in other Dok family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| WB, E | |
| Unconjugated | |
| DOK1 | |
| AW557123; Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); Dok; DOK 1; DOK1; Dok-1; Downstream of tyrosine kinase 1; downstream of tyrosine kinase-1; MGC117395; MGC138860; p240; p62 dok; p62(dok); p62DOK; pp62; TLP1; TP1; TROVE domain family; TROVE1; VAULT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13448, 1796, 312477 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction