missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DNA-PK (Ser2056) Recombinant Rabbit Monoclonal Antibody (23GB2075), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553214
This item is not returnable.
View return policy
Description
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
Specifications
| Phospho-DNA-PK (Ser2056) | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRKDC | |
| AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5591 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2075 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P78527 | |
| PRKDC | |
| A synthesized peptide derived from human Phospho-DNA PKcs (Ser2056) (2049-2063AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction