missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DEPDC6 (Ser244) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105064
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of DEPTOR only when phosphorylated at Ser244.
DEPDC6 is a gene that encodes a protein known for its role in various cellular processes, although the specifics of its function remain not fully elucidated. Located on chromosome 22, DEPDC6 is involved in regulating cell growth and survival by modulating the RAS signaling pathway, a crucial pathway for many cellular functions. Structurally, DEPDC6 contains a DEP domain, which is known to mediate interactions with other proteins that are involved in signal transduction. This domain plays a significant role in facilitating the proper localization and function of the protein. Functionally, DEPDC6 is implicated in controlling the pathways that govern the timing and progression of the cell cycle, highlighting its importance in maintaining cellular homeostasis and preventing uncontrolled cell proliferation. Its dysregulation has been associated with various cancers, emphasizing the need for further studies to fully understand its potential as a therapeutic target.
Specifications
| Phospho-DEPDC6 (Ser244) | |
| Polyclonal | |
| Unconjugated | |
| DEPTOR | |
| DEP domain containing 6; DEP domain containing MTOR interacting protein; DEP domain containing MTOR-interacting protein; DEP domain-containing mTOR-interacting protein; DEP domain-containing protein 6; DEP.6; Depdc6; DEPTOR; Kiaa4200; RGD1561030 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 314979, 64798, 97998 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q570Y9, Q8TB45 | |
| DEPTOR | |
| A synthesized peptide derived from human DEPTOR(Accession Q8TB45), corresponding to amino acid residues around phosphorylated Ser244. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction