missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dematin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599467
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Dematin.
The protein encoded by this gene is an actin binding and bundling protein that plays a structural role in erythrocytes, by stabilizing and attaching the spectrin/actin cytoskeleton to the erythrocyte membrane in a phosphorylation-dependent manner. This protein contains a core domain in the N-terminus, and a headpiece domain in the C-terminus that binds F-actin. When purified from erythrocytes, this protein exists as a trimer composed of two 48 kDa polypeptides and a 52 kDa polypeptide. The different subunits arise from alternative splicing in the 3' coding region, where the headpiece domain is located. Disruption of this gene has been correlated with the autosomal dominant Marie Unna hereditary hypotrichosis disease, while loss of heterozygosity of this gene is thought to play a role in prostate cancer progression. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| Dematin | |
| Polyclonal | |
| Unconjugated | |
| DMTN | |
| AI325486; dematin; dematin actin binding protein; dematin actin-binding protein; DMT; Dmtn; Epb4.9; Epb49; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin); erythrocyte protein band 4.9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13829, 2039 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q08495, Q9WV69 | |
| DMTN | |
| A synthesized peptide derived from human DMTN(Accession Q08495), corresponding to amino acid residues S377-F405. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction