missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DARPP-32 (Thr75) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14611
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, chicken, human, mouse and non-human primate based on 100% sequence homology. In Western blot, this antibody detects an ~32 kDa protein representing the Phospho-Thr75 DARPP-32 protein.
DARPP-32 is a dopamine (DA) and AMP-regugated ~32 kDa phosphoprotein that is associated with dopaminoceptive neurons. The protein inhibits Protein Phosphatase I when it is phosphorylated on Thr34. In contrast, when DARPP-32 is phosphorylated on Thr75 the protein acts as an inhibitor of PKA. Phosphorylation of DARPP-32 is thought to play a critical role in the regugation of dopaminergic neurotransmission. In addition, the activity of DARPP-32 is also thought to play important roles in the actions of alcohol, caffeine and Prozac™.
Specifications
| Phospho-DARPP-32 (Thr75) | |
| Polyclonal | |
| Unconjugated | |
| Ppp1r1b | |
| AU040756; DARPP32; DARPP-32; dopamine- and cAMP-regulated neuronal phosphoprotein; dopamine and cAMP-regulated neuronal phosphoprotein 32; dopamine- and cAMP-regulated phosphoprotein DARPP-32; FLJ20940; IPPD; neuronal phosphoprotein DARPP-32; OTTHUMP00000164275; OTTHUMP00000164276; Ppp1r1b; PPR1B; protein pho; protein phosphatase 1 regulatory inhibitor subunit 1B; protein phosphatase 1 regulatory subunit 1B; protein phosphatase 1, regulatory (inhibitor) subunit 1B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19049, 360616 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q60829, Q6J4I0 | |
| Ppp1r1b | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr75 of rat PPP1R1B conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction