missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DAB1 (Tyr232) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104709
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Dab1 only when phosphorylated at Tyrosine 232.
The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
Specifications
| Phospho-DAB1 (Tyr232) | |
| Polyclonal | |
| Unconjugated | |
| DAB1 | |
| AI956902; C630028C02Rik; Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Dab, reelin signal transducer, homolog 1 (Drosophila); Dab1; DAB1, reelin adaptor protein; disabled 1; disabled homolog 1; RP23-356E21.1; scm; scr; scrambler; yot; yotari | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 13131, 1600, 266729 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75553, P97318, Q8CJH2 | |
| DAB1 | |
| A synthesized peptide derived from human DAB1(Accession O75553), corresponding to amino acid residues around phosphorylated Tyr232. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction