missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Cyclin D2 (Thr280) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104257
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Cyclin D2 only when phosphorylated at Thr280.
Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
Specifications
| Phospho-Cyclin D2 (Thr280) | |
| Polyclonal | |
| Unconjugated | |
| CCND2 | |
| 2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 12444, 64033, 894 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30279, P30280, Q04827 | |
| CCND2 | |
| A synthesized peptide derived from human CCND2(Accession P30279), corresponding to amino acid residues around phosphorylated Thr280. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction