missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-CrkL (Tyr207) Recombinant Rabbit Monoclonal Antibody (CrkLY207-G4), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£185.00 - £463.00
Specifications
| Antigen | Phospho-CrkL (Tyr207) |
|---|---|
| Clone | CrkLY207-G4 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17166848
|
Invitrogen™
MA537260 |
10 Tests |
£185.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17186558
![]() |
Invitrogen™
MA536972 |
100 Tests |
£463.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May mediate the transduction of intracellular signals.Specifications
| Phospho-CrkL (Tyr207) | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Human, Mouse | |
| P46109, P47941 | |
| 12929, 1399 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr207 of human phospho CrkL. | |
| Primary | |
| 4°C | |
| CRKL |
| CrkLY207-G4 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 1110025F07Rik; AA589403; AI325100; CRK like proto-oncogene, adaptor protein; CRKL; crkl {ECO:0000312; crk-like protein; crk-like protein {ECO:0000250; Crkol; EMBL:AAH85865.1, ECO:0000312; OTTHUMP00000198539; RGD:1308531}; UniProtKB:P46109}; v-crk avian sarcoma virus CT10 oncogene homolog-like; v-crk sarcoma virus CT10 oncogene homolog (avian)-like; v-crk sarcoma virus CT10 oncogene homolog-like | |
| CRKL | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title