missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Creatine Kinase MT (Tyr153) Polyclonal Antibody

Product Code. 16528521
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
16528521 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 16528521

Brand: Invitrogen PA595676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phospho-Creatine Kinase MT (Tyr153) Polyclonal Antibody

Phospho-Creatine Kinase MT (Tyr153) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Creatine Kinase MT (Tyr153)
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene CKMT1A
Gene Accession No. P12532, P30275
Gene Alias acidic-type mitochondrial creatine kinase; CKMT; CKMT1; CKMT1A; CKMT1B; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); creatine kinase, mitochondrial 1, ubiquitous; creatine kinase, mitochondrial 1A; creatine kinase, mitochondrial 1B; mia-CK; mi-CK; Mt-CK; ScCKmit; UbCKmit; ubiquitous mitochondrial creatine kinase; UMTCK; U-MtCK
Gene Symbols Ckmt1, CKMT1A, CKMT1B
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr153 conjugated to KLH.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1159, 12716, 548596
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.