missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Cofilin (Ser3) Monoclonal Antibody (G.577.4)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA515038
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.
Specifications
| Phospho-Cofilin (Ser3) | |
| Monoclonal | |
| 42 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| P18760, P23528, P45592, Q5E9F7 | |
| CFL1 | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer3 of human cofilin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| G.577.4 | |
| Unconjugated | |
| CFL1 | |
| 18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1072, 12631, 29271, 534553 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction