missing translation for 'onlineSavingsMsg'
Learn More

Phospho-CK2 alpha-1 (Tyr255) Polyclonal Antibody, Invitrogen™

Product Code. 15575741
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15575741 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15575741 Supplier Invitrogen Supplier No. PA538831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Casein kinase II is a serine/threonine protein kinase that phosphorylates acidic proteins such as casein. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by this gene represents the alpha subunit. While this gene is found on chromosome 20, a related transcribed pseudogene is found on chromosome 11. Three transcript variants encoding two different proteins have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CK2 alpha-1 (Tyr255)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSNK2A1
Gene Accession No. P68400, Q60737, P19139
Gene Alias CK2A1, CKII, CK II alpha, CK2 catalytic subunit alpha, casein kinase II alpha 1 subunit, casein kinase II subunit alpha, protein kinase CK2
Gene Symbols CSNK2A1
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human Casein Kinase II alpha around the phosphorylation site of Tyr255 (D-L-YP-D-Y)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116549, 12995, 1457
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.