missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-CHK1 (Ser345) Monoclonal Antibody (S.48.4)

Description
It is not recommended to aliquot this antibody.
CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.
Specifications
Specifications
| Antigen | Phospho-CHK1 (Ser345) |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | S.48.4 |
| Concentration | 37 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100 μg/mL BSA and <0.02% sodium azide; pH 7.5 |
| Gene | Chek1 |
| Gene Accession No. | O14757, O35280, Q91ZN7 |
| Gene Alias | C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?