missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CHEK2 (Thr68) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£109.00 - £249.00
Specifications
| Antigen | CHEK2 (Thr68) |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30238041
|
Proteintech
29012-1-AP-100UL |
100 μL |
£280.00 £249.00 / 100µL Save £31.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236166
|
Proteintech
29012-1-AP-20UL |
20 μL |
£125.00 £109.00 / 20µL Save £16.00 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-CHEK2 (Thr68) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-CHEK2 (Thr68). This antibody recognizes human antigen. The Phospho-CHEK2 (Thr68) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| CHEK2 (Thr68) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CDS1; CHK2; HuCds1; LFS2; PP1425; RAD53 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 450 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 11200 | |
| Peptide | |
| Primary | |
| Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title