missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-CDK9 (Thr186) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517197
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
CDK9 is a serine/threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle. CDK9 is coexpressed and copurified with CyclinT1. CDK9 regulates cytokine inducible transcription networks by facilitating promoter recognition of target transcription factors. CDK9 promotes RNA synthesis in genetic programs for cell growth, differentiation, and viral pathogenesis. The CDK9/cyclin-K complex also has kinase activity towards CTD of RNAPII and can substitute for CDK9/cyclin-T P-TEFb in vitro. Chronic activation of CDK9 causes cardiac myocyte enlargement leading to cardiac hypertrophy and confers a predisposition to heart failure.
Specifications
| Phospho-CDK9 (Thr186) | |
| Polyclonal | |
| Unconjugated | |
| CDK9 | |
| C-2k; CDC2L4; CDC2-related kinase; Cdk9; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; CTK1; cyclin dependent kinase 9; cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); PITALRE; RP11-228B15.5; serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; TAK; Tat-associated kinase complex catalytic subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1025, 107951, 362110 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 126 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P50750, Q641Z4, Q99J95 | |
| CDK9 | |
| Synthetic phosphopeptide corresponding to residues surrounding pThr186 of human CDK9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction