missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-CDK1/CDK2 (Thr14) Recombinant Rabbit Monoclonal Antibody (K01_2K81)

Product Code. 30830288
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830288 50 μL
30827906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830288 Supplier Invitrogen™ Supplier No. MA565431
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-CDK1/CDK2 (Thr14) Recombinant Monoclonal Antibody for IHC (P), Western Blot

Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CDK1/CDK2 (Thr14)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2K81
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CDK2
Gene Accession No. P06493, P24941
Gene Alias A630093N05Rik; Cdc2; CDC28A; Cdc2a; cdc2-related protein kinase; Cdk1; CDK2; CDKN1; CDKN2; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control protein 2 homolog; cell division cycle 2 homolog A; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 2; cyclin dependent kinase 1; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 1; Cyclin-dependent kinase 2; p33 protein kinase; p33(CDK2); p34 protein kinase; p34>CDC2>; P34CDC2
Gene Symbols CDK1, CDK2
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Thr14 of human Cdk1/2.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1017, 983
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.