missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Cdc6 (Ser106) Recombinant Rabbit Monoclonal Antibody (K01_3L22)

Product Code. 30824728
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824728 100 μL
30829595 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824728 Supplier Invitrogen™ Supplier No. MA564383
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-Cdc6 (Ser106) Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Cdc6 (Ser106)
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3L22
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CDC6
Gene Accession No. Q99741
Gene Alias CDC18(S.pombe); CDC18L; cdc18-related protein; CDC6; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6; cell division cycle 6 homolog; HsCDC18; HsCDC6; p62(cdc6)
Gene Symbols CDC6
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser106 of human Cdc6.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360621, 990
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.