missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Phospho-Cdc25A (Ser178) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104863
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Antibody detects endogenous levels of CDC25A only when phosphorylated at Serine 178.
CDC25a is a member of the CDC25 family of phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. CDC25a is required for progression through G1 and S phases of the cell cycle. CDC25a activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division. CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated.
Tekniske data
| Phospho-Cdc25A (Ser178) | |
| Polyclonal | |
| Unconjugated | |
| CDC25A | |
| CDC25 isoform A1-CAG; CDC25A; CDC25A2; CDC25A2-CAG isoform; Cdc25m3; cell division cycle 25A; D9Ertd393e; dual specificity phosphatase Cdc25A; EC 3.1.3.48; M-phase inducer phosphatase 1; MPIP1 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 12530, 171102, 993 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30304, P48964, P48965 | |
| CDC25A | |
| A synthesized peptide derived from human CDC25A(Accession P30304), corresponding to amino acid residues around phosphorylated Ser178. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion