Learn More
Invitrogen™ Phospho-CD247 (CD3 zeta) (Tyr142) Monoclonal Antibody (3ZBR4S), PE, eBioscience™, Invitrogen™

Description
Description: This 3ZBR4S monoclonal antibody recognizes human and mouse CD247 (CD3z) when phosphorylated on Tyrosine 142. CD3 is a multi-protein complex expressed on the surface of thymocytes and T cells, a marker of the T cell lineage, and a critical component of T cell receptor (TCR) signaling. The complex contains a gamma chain, a delta chain, and two epsilon chains, which associate with the alpha/beta or gamma/delta chains of the TCR, and the zeta chain homodimer. The CD3 zeta chain is responsible for initiating the signaling cascade following engagement of the TCR. Each CD3 zeta chain contains three immunoreceptor tyrosine-based activation motifs (ITAM) that are phosphorylated by Src kinases such as Lck. Once phosphorylated, these ITAMs form docking sites for SH2 domain-containing proteins and function to recruit ZAP-70 to the TCR where it can further amplify and propagate signaling. Loss or mutation of CD3 zeta or the ITAM motifs results in impaired T cell development, signaling, and function. CD3 zeta phosphorylation occurs early in the TCR signaling cascade. It occurs immediately downstream of Lck activation, and upstream of ZAP-70, LAT, and SLP-76. Applications Reported: This 3ZBR4S antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3ZBR4S antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test.
Specifications
Specifications
| Antigen | Phospho-CD247 (CD3 zeta) (Tyr142) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3ZBR4S |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD247 |
| Gene Accession No. | P20963, P24161 |
| Gene Alias | 4930549J05Rik; A430104F18Rik; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; Cd3; CD3 antigen, zeta polypeptide; CD3 zeta; CD3 zeta chain; CD3e antigen; CD3-epsilon; Cd3-eta; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; FLJ18683; IMD25; T3E; T3z; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 eta chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; TCRZ; TCRzeta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.