missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-CCR5 (Ser349) Chimeric Recombinant Rabbit Monoclonal Antibody (E11/19)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548293
This item is not returnable.
View return policy
Description
Specificity: The E11/19 monoclonal antibody binds to phosphoserine 349 corresponding to 22 c-terminal amino acid residue of human CCR5. CCR5 is a receptor for a number of inflammatory CC-chemokines including CCL3/MIP-1-alpha, CCL4/MIP-1-beta and RANTES.
Human immunodeficiency virus (HIV) and related virus require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CCR5 (CC-CKR-5) is a principal coreceptor for macrophage- and dual-tropic HIV-1 strains fusion and entry of human white blood cells. CCR5 is required for the infection by HIV-1, HIV-2, and SIV. The beta-chemokines RANTES, MIP-alpha and MIP-beta are the ligands for CCR5 and prevent infection by M-tropic HIV-1. CCR5 associates with the surface CD4-gp120 of HIV complex and leads to membrane fusion and virus entry of target cells. The amino-terminal domain and the extracellular loops of CCR5 serve as HIV biding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.
Specifications
| Phospho-CCR5 (Ser349) Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P51681 | |
| CCR5 | |
| This antibody was raised by immunizing mice with a synthetic phosphopeptide (CEAPERA(pS)(pS)VYTR(pS)TGEQI(pS)VGL) corresponding to the C-terminal region of human CCR5. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| E11/19 | |
| Unconjugated | |
| CCR5 | |
| AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1234 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction