missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CaMKI (Thr177) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538434
This item is not returnable.
View return policy
Description
Calcium/calmodulin-dependent protein kinase I is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Specifications
| Phospho-CaMKI (Thr177) | |
| Polyclonal | |
| Unconjugated | |
| CAMK1 | |
| CAMKI, caM kinase I alpha, caM-KI, caMKI-alpha, calcium/calmodulin-dependent protein kinase type 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171503, 52163, 8536 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14012, Q91YS8, Q63450 | |
| CAMK1 | |
| A synthetic phosphopeptide derived from human CaMK1-alpha around the phosphorylation site of Thr177 (L-S-TP-A-C) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction