missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-c-Raf (Ser259) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 44502
This item is not returnable.
View return policy
Description
Phospho-c-Raf (Ser259) Polyclonal Antibody for Western Blot, ICC/IF, IP
Raf-1 (c-Raf) is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated Raf-1 can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylate to activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2.
Specifications
| Phospho-c-Raf (Ser259) | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
| P04049 | |
| Raf1 | |
| antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human c-Raf that contains serine 259. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Raf1 | |
| 6430402F14Rik; AA990557; BB129353; CMD1NN; CRAF; C-RAF; C-Raf proto-oncogene, serine/threonine kinase; Craf1; Craf1 transforming; D830050J10Rik; EC 2.7.11.1; kinase Raf1; murine leukemia viral (v-raf-1) oncogene homolog 1 (3611-MSV); murine sarcoma 3611 oncogene 1; NS5; Oncogene RAF1; OTTHUMP00000207813; OTTHUMP00000209389; proto-oncogene c-RAF; RAF; raf proto-oncogene serine/threonine protein kinase; RAF proto-oncogene serine/threonine-protein kinase; RAF1; Raf-1; Raf-1 proto-oncogene, serine/threonine kinase; sarcoma 3611 oncogene; v-Raf; v-raf-1 leukemia viral oncogene 1; vraf1 murine le; v-raf-1 murine leukemia viral oncogene homolog 1; v-raf-1 murine leukemia viral oncogene-like protein 1; v-raf-leukemia viral oncogene 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5894 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction