Learn More
Phospho-c-MYC (Ser62) Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28915-1-AP-20UL
Description
The Phospho-c-MYC (Ser62) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-c-MYC (Ser62). This antibody recognizes human antigen. The Phospho-c-MYC (Ser62) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-c-MYC (Ser62) | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| bHLHe39, c Myc, MRTL, MYC, Myc proto oncogene protein, Phopho-Ser62-MYC, Proto oncogene c Myc, Transcription factor p64 | |
| 20 μL | |
| 4609 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |