missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-c-Jun (Tyr170) Polyclonal Antibody

Phospho-c-Jun (Tyr170) Polyclonal Antibody
Brand: Invitrogen PA536634
This item is not returnable.
View return policy
Description
Phospho-c-Jun (Tyr170) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Phospho-c-Jun (Tyr170) | |
| Polyclonal | |
| Unconjugated | |
| JUN | |
| Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16476, 24516, 3725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05412, P05627, P17325 | |
| JUN | |
| Synthetic phosphopeptide derived from human c-Jun around the phosphorylation site of Tyrosine 170. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction