missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-c-Jun (Ser73) Recombinant Rabbit Monoclonal Antibody (HL2713)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556650
This item is not returnable.
View return policy
Description
Phospho-c-Jun (Ser73) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
Specifications
| Phospho-c-Jun (Ser73) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P05412 | |
| JUN | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Ser73 of human c-Jun. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2713 | |
| Unconjugated | |
| JUN | |
| Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3725 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction