missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-C/EBP epsilon (Ser181) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105657
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of C/EBP epsilon only when phosphorylated at ser181.
C/EBP alpha regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP alpha uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic alpha helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP alpha appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP beta, C/EBP gamma, C/EBP delta and C/EBP epsilon, all of which exhibit similar DNA-binding specificities and affinities to C/EBP alpha. Furthermore, C/EBP beta and C/EBP delta readily form heterodimers both with each other as well as with C/EBP alpha.
Specifications
| Phospho-C/EBP epsilon (Ser181) | |
| Polyclonal | |
| Unconjugated | |
| CEBPE | |
| C EBP; c/EBP epsilon; C/EBPe; C/EBPepsilon; C/EBP-epsilon; C/EBP-related protein 1; C/EPBe; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein epsilon; CEBPE; Crp1; Gm294 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 1053, 110794, 25410 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56261, Q15744, Q6PZD9 | |
| CEBPE | |
| A synthesized peptide derived from human CEBPE(Accession Q15744), corresponding to amino acid residues around phosphorylated Ser181. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction