missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-C/EBP alpha (Thr230) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538155
This item is not returnable.
View return policy
Description
The C/EBP (CCAAT/enhancer binding protein) family of transcription factors--alpha, beta, delta, and epsilon)--are widely expressed proteins found in a variety of tissues. These domains contain an evolutionarily conserved five-amino acid motif (the regulatory domain motif RDM) that conforms to the consensus sequence (I/V/L) KXEP. One of the major roles of these transcription factors is that of metabolic regulation in liver and fat cells. Studies suggest that these transcription factors act in a cascade aiding in the development of the adipose cell. Three C/EBP beta isoforms (LAP*, LAP, and LIP) arise from different translation initiation and display different function in gene regulation. From a neurological standpoint, C/EBPs have been implicated in the mediation of nerve growth factor, altering synaptic plasticity, and mediation of inflammatory response in AD patients.
Specifications
| Phospho-C/EBP alpha (Thr230) | |
| Polyclonal | |
| Unconjugated | |
| CEBPA | |
| CCAAT/enhancer-binding protein alpha C/EBP alpha CEBPA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1050, 12606, 24252 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49715, P53566, P05554 | |
| CEBPA | |
| A synthetic phosphopeptide derived from human C/EBP-alpha around the phosphorylation site of Thr230 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction